Gene Transcripts
- Gene Symbol
- AHCYL1
- Entrez Gene ID
- 10768
- Gene Name
- adenosylhomocysteinase like 1
- Gene Aliases
- DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA
- Chromosome Location
- Chr. 1: 109984741 - 110023742 on Build GRCh38
- UniGene ID
- Hs.743973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF090905.1 |
- |
1 |
169 |
|
AK131563.1 |
- |
10 |
2083 |
|
AK313392.1 |
- |
1 |
411 |
|
View Full Product Details