Gene Transcripts
- Gene Symbol
- CDK13
- Entrez Gene ID
- 8621
- Gene Name
- cyclin dependent kinase 13
- Gene Aliases
- CDC2L, CDC2L5, CHED, hCDK13
- Chromosome Location
- Chr. 7: 39949501 - 40097134 on Build GRCh38
- UniGene ID
- Hs.233552
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130653.1 |
- |
1 |
251 |
|
View Full Product Details