Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011519387.2 | XP_011517689.1 | 1 | 1938 | |
XM_017015909.1 | XP_016871398.1 | 1 | 1938 | ||
XM_017015910.1 | XP_016871399.1 | 1 | 1938 | ||
XM_017015911.1 | XP_016871400.1 | 1 | 1937 | ||
XM_017015913.1 | XP_016871402.1 | 1 | 1937 | ||
XM_017015914.1 | XP_016871403.1 | 1 | 1937 | ||
GenBank mRNA | BC047402.1 | - | 2 | 770 |