Gene Transcripts
- Gene Symbol
- TCL6
- Entrez Gene ID
- 27004
- Gene Name
- T-cell leukemia/lymphoma 6 (non-protein coding)
- Gene Aliases
- TNG1, TNG2
- Chromosome Location
- Chr. 14: 95651178 - 95673452 on Build GRCh38
- UniGene ID
- Hs.510368
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023714.1 |
- |
1 |
73 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW504203.1 |
- |
1 |
308 |
|
Chr. 14 - 95675919 - 95676279 [+] |
GenBankmRNA |
AU139853.1 |
- |
1 |
73 |
|
Chr. 14 - 95676152 - 95676905 [+] |
View Full Product Details