Gene Transcripts
- Gene Symbol
- CDKAL1
- Entrez Gene ID
- 54901
- Gene Name
- CDK5 regulatory subunit associated protein 1 like 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 20534457 - 21232404 on Build GRCh38
- UniGene ID
- Hs.657604
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074342.1 |
- |
1 |
1129 |
|
View Full Product Details