Gene Transcripts
- Gene Symbol
- CMTM1
- Entrez Gene ID
- 113540
- Gene Name
- CKLF like MARVEL transmembrane domain containing 1
- Gene Aliases
- CKLFH, CKLFH1, CKLFSF1
- Chromosome Location
- Chr. 16: 66566391 - 66579135 on Build GRCh38
- UniGene ID
- Hs.15159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC018586.1 |
- |
1 |
1416 |
|
BC028676.1 |
- |
1 |
484 |
|
- Gene Symbol
- CKLF
- Entrez Gene ID
- 51192
- Gene Name
- chemokine-like factor
- Gene Aliases
- C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1
- Chromosome Location
- Chr. 16: 66552563 - 66566287 on Build GRCh38
- UniGene ID
- Hs.15159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK301240.1 |
- |
5 |
1051 |
|
View Full Product Details