Gene Transcripts
- Gene Symbol
- Poglut1
- Entrez Gene ID
- 224143
- Gene Name
- protein O-glucosyltransferase 1
- Gene Aliases
- 9630046K23Rik, Clp46, Ktelc1, Rumi
- Chromosome Location
- Chr. 16: 38525058 - 38550256 on Build GRCm38
- UniGene ID
- Mm.284366
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076379.1 |
- |
|
2198 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB007121.2 |
- |
1 |
546 |
|
Chr. 16 - 38523197 - 38523859 [-] |
View Full Product Details