Gene Transcripts
- Gene Symbol
- Dtna
- Entrez Gene ID
- 13527
- Gene Name
- dystrobrevin alpha
- Gene Aliases
- 2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn
- Chromosome Location
- Chr. 18: 23310020 - 23659719 on Build GRCm38
- UniGene ID
- Mm.484047
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008850.1 |
- |
1 |
320 |
|
AK012743.1 |
- |
1 |
320 |
|
AK134543.1 |
- |
1 |
575 |
|
View Full Product Details