Gene Transcripts
- Gene Symbol
- Igsf3
- Entrez Gene ID
- 78908
- Gene Name
- immunoglobulin superfamily, member 3
- Gene Aliases
- 1700016K10Rik, 2810035F16Rik, 4833439O17Rik, mKIAA0466
- Chromosome Location
- Chr. 3: 101377011 - 101463060 on Build GRCm38
- UniGene ID
- Mm.257997
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006042.1 |
- |
1, 2 |
106 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706159.1 |
- |
1, 2 |
103 |
|
Chr. 3 - 101425274 - 101426936 [-] |
View Full Product Details