Gene Transcripts
- Gene Symbol
- Rapgef5
- Entrez Gene ID
- 217944
- Gene Name
- Rap guanine nucleotide exchange factor (GEF) 5
- Gene Aliases
- 4932413M22, C86120, D030051B22Rik, GFR, Mrgef, mKIAA0277, mmr-gef
- Chromosome Location
- Chr. 12: 117516479 - 117759737 on Build GRCm38
- UniGene ID
- Mm.227642
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK049664.1 |
- |
1 |
2515 |
|
View Full Product Details