Gene Transcripts
- Gene Symbol
- Pax5
- Entrez Gene ID
- 18507
- Gene Name
- paired box 5
- Gene Aliases
- BSAP, EBB-1, KLP, Pax-5
- Chromosome Location
- Chr. 4: 44526269 - 44710481 on Build GRCm38
- UniGene ID
- Mm.439659
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK144818.1 |
- |
1 |
2646 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB817085.1 |
- |
1 |
267 |
|
Chr. 4 - 44526386 - 44526781 [-] |
View Full Product Details