Gene Transcripts
- Gene Symbol
- Vsig10l
- Entrez Gene ID
- 75690
- Gene Name
- ZV-set and immunoglobulin domain containing 10 like
- Gene Aliases
- Chromosome Location
- Chr. 7: 43463061 - 43472023 on Build GRCm38
- UniGene ID
- Mm.183297
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008908.1 |
- |
1 |
473 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE626902.1 |
- |
1 |
175 |
|
Chr. 7 - 43471355 - 43471913 [+] |
View Full Product Details