Gene Transcripts
- Gene Symbol
- Nsun3
- Entrez Gene ID
- 106338
- Gene Name
- NOL1/NOP2/Sun domain family member 3
- Gene Aliases
- 6720484A09Rik, AU022521
- Chromosome Location
- Chr. 16: 62732444 - 62786794 on Build GRCm38
- UniGene ID
- Mm.26321
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020172.1 |
- |
1 |
859 |
|
AK088718.1 |
- |
1 |
859 |
|
AK162988.1 |
- |
1 |
850 |
|
View Full Product Details