Gene Transcripts
- Gene Symbol
- Vps13b
- Entrez Gene ID
- 666173
- Gene Name
- vacuolar protein sorting 13B (yeast)
- Gene Aliases
- 1810042B05Rik, 2310042E16Rik, 4732488H20, C330002D13Rik, C87206, Coh1, D230005K13, mKIAA0532, mKIAA5032
- Chromosome Location
- Chr. 15: 35369882 - 35931229 on Build GRCm38
- UniGene ID
- Mm.309250
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK051828.1 |
- |
1 |
2744 |
|
View Full Product Details