Gene Transcripts
- Gene Symbol
- Ttll13
- Entrez Gene ID
- 269954
- Gene Name
- tubulin tyrosine ligase-like family, member 13
- Gene Aliases
- 1700111A04Rik, 4921538O11, BC024578
- Chromosome Location
- Chr. 7: 80246376 - 80260821 on Build GRCm38
- UniGene ID
- Mm.276780
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007167.1 |
- |
1 |
292 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707251.1 |
- |
1 |
292 |
|
Chr. 7 - 80246425 - 80246806 [+] |
View Full Product Details