Gene Transcripts
- Gene Symbol
- Zim3
- Entrez Gene ID
- 116811
- Gene Name
- zinc finger, imprinted 3
- Gene Aliases
- 1700128I23Rik
- Chromosome Location
- Chr. 7: 6955685 - 6976662 on Build GRCm38
- UniGene ID
- Mm.381671
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007312.2 |
- |
|
194 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707418.1 |
- |
1, 2 |
194 |
|
Chr. 7 - 6976302 - 6977420 [-] |
View Full Product Details