Gene Transcripts
- Gene Symbol
- Ttc7
- Entrez Gene ID
- 225049
- Gene Name
- tetratricopeptide repeat domain 7
- Gene Aliases
- 1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
- Chromosome Location
- Chr. 17: 87282082 - 87381770 on Build GRCm38
- UniGene ID
- Mm.77396
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005733.1 |
- |
1 |
75 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705898.1 |
- |
1 |
70 |
|
Chr. 17 - 87329789 - 87332049 [-] |
View Full Product Details