Gene Transcripts
- Gene Symbol
- Dcdc5
- Entrez Gene ID
- 329482
- Gene Name
- doublecortin domain containing 5
- Gene Aliases
- 4732421G10Rik, BC048594, EG436559
- Chromosome Location
- Chr. 2: 106042135 - 106408603 on Build GRCm38
- UniGene ID
- Mm.389384
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138215.1 |
- |
3 |
313 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY723817.1 |
- |
3 |
313 |
|
Chr. 2 - 106226454 - 106336145 [+] |
View Full Product Details