Gene Transcripts
- Gene Symbol
- Kcnn4
- Entrez Gene ID
- 16534
- Gene Name
- potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4
- Gene Aliases
- IK1, IKCA1, KCA4, KCa3.1, SK4, SKCas, mIKCa1
- Chromosome Location
- Chr. 7: 24369833 - 24385212 on Build GRCm38
- UniGene ID
- Mm.9911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038253.1 |
- |
|
3619 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB166842.2 |
- |
1 |
619 |
|
Chr. 7 - 24383054 - 24383686 [-] |
View Full Product Details