Gene Transcripts
- Gene Symbol
- Ccdc37
- Entrez Gene ID
- 243538
- Gene Name
- coiled-coil domain containing 37
- Gene Aliases
- AW048498, C030041G11Rik, C230069K22Rik
- Chromosome Location
- Chr. 6: 90403731 - 90430246 on Build GRCm38
- UniGene ID
- Mm.386876
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021139.1 |
- |
2, 3 |
219 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM936873.1 |
- |
2, 3 |
535 |
|
Chr. 6 - 90421230 - 90428797 [-] |
GenBankmRNA |
BY728284.1 |
- |
2, 3 |
219 |
|
Chr. 6 - 90420938 - 90428480 [-] |
GenBankmRNA |
CA466194.1 |
- |
|
176 |
|
Chr. - - [-] |
View Full Product Details