Gene Transcripts
- Gene Symbol
- Adgrl3
- Entrez Gene ID
- 319387
- Gene Name
- adhesion G protein-coupled receptor L3
- Gene Aliases
- 5430402I23Rik, CIRL-3, D130075K09Rik, Gm1379, LEC3, Lphn3, mKIAA0768
- Chromosome Location
- Chr. 5: 81021199 - 81827825 on Build GRCm38
- UniGene ID
- Mm.273631
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017260.1 |
- |
1 |
934 |
|
AK045510.1 |
- |
1 |
364 |
|
AK122367.1 |
- |
18 |
4294 |
|
View Full Product Details