Gene Transcripts
- Gene Symbol
- Galr2
- Entrez Gene ID
- 14428
- Gene Name
- galanin receptor 2
- Gene Aliases
- mGalR
- Chromosome Location
- Chr. 11: 116280232 - 116287498 on Build GRCm38
- UniGene ID
- Mm.57149
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK137830.1 |
- |
1 |
2174 |
|
View Full Product Details