Gene Transcripts

Gene Symbol
THTPA
Entrez Gene ID
79178
Gene Name
thiamine triphosphatase
Gene Aliases
THTP, THTPASE
Chromosome Location
Chr. 14: 23511760 - 23559581 on Build GRCh38
UniGene ID
Hs.655179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK094211.1 - 3 2074


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000553985.1 - 3 2074 Chr. 14 - 23512817 - 23560778 [+]

View Full Product Details