Gene Transcripts

Gene Symbol
PPIEL
Entrez Gene ID
728448
Gene Name
peptidylprolyl isomerase E like pseudogene
Gene Aliases
PPIEP1
Chromosome Location
Chr. 1: 39522280 - 39559698 on Build GRCh38
UniGene ID
Hs.472508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK093659.1 - 2 1956


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000441741.5 - 3 208 Chr. 1 - 39522280 - 39524876 [-]

View Full Product Details