Gene Transcripts
- Gene Symbol
- GNAS-AS1
- Entrez Gene ID
- 149775
- Gene Name
- GNAS antisense RNA 1
- Gene Aliases
- GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
- Chromosome Location
- Chr. 20: 58818918 - 58850903 on Build GRCh38
- UniGene ID
- Hs.122718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002785.2 |
|
5 |
1112 |
|
GenBank mRNA |
AJ251759.1 |
- |
5 |
1112 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000424094.6 |
- |
5 |
1112 |
|
Chr. 20 - 58818919 - 58850903 [-] |
View Full Product Details