Gene Transcripts
- Gene Symbol
- TNFRSF10B
- Entrez Gene ID
- 8795
- Gene Name
- TNF receptor superfamily member 10b
- Gene Aliases
- CD262, DR5, KILLER, KILLER/DR5, TRAIL-R2, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9
- Chromosome Location
- Chr. 8: 23020133 - 23069187 on Build GRCh38
- UniGene ID
- Hs.521456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC043400.1 |
- |
2 |
852 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000502083.2 |
- |
2 |
852 |
|
Chr. 8 - 23006381 - 23019335 [-] |
View Full Product Details