Gene Transcripts
- Gene Symbol
- RIMBP2
- Entrez Gene ID
- 23504
- Gene Name
- RIMS binding protein 2
- Gene Aliases
- PPP1R133, RBP2, RIM-BP2
- Chromosome Location
- Chr. 12: 130396133 - 130716361 on Build GRCh38
- UniGene ID
- Hs.657441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX641152.1 |
- |
1 |
1895 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000539532.1 |
- |
2 |
175 |
|
Chr. 12 - 130419535 - 130421019 [-] |
View Full Product Details