Gene Transcripts
- Gene Symbol
- PRC1-AS1
- Entrez Gene ID
- 100507118
- Gene Name
- PRC1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 90966369 - 90988624 on Build GRCh38
- UniGene ID
- Hs.366401
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_051984.1 |
|
4 |
1036 |
|
GenBank mRNA |
AA403001.1 |
- |
1 |
185 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000554388.1 |
- |
4 |
1033 |
|
Chr. 15 - 90966372 - 90988624 [+] |
View Full Product Details