Gene Transcripts
- Gene Symbol
- PRC1-AS1
- Entrez Gene ID
- 100507118
- Gene Name
- PRC1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 90966369 - 90988624 on Build GRCh38
- UniGene ID
- Hs.366401
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_051984.1 |
|
4 |
765 |
|
GenBank mRNA |
BI829394.1 |
- |
4 |
737 |
|
CD244496.1 |
- |
4 |
790 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI829185.1 |
- |
4 |
746 |
|
Chr. 15 - 90966388 - 90987917 [+] |
GENCODE |
ENST00000554388.1 |
- |
4 |
762 |
|
Chr. 15 - 90966372 - 90988624 [+] |
View Full Product Details