Gene Transcripts
- Gene Symbol
- CHMP4A
- Entrez Gene ID
- 29082
- Gene Name
- charged multivesicular body protein 4A
- Gene Aliases
- C14orf123, CHMP4, CHMP4B, HSPC134, SHAX2, SNF7, SNF7-1, VPS32-1, VPS32A
- Chromosome Location
- Chr. 14: 24209581 - 24213830 on Build GRCh38
- UniGene ID
- Hs.279761
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM041056.1 |
- |
1 |
66 |
|
- Gene Symbol
- MDP1
- Entrez Gene ID
- 145553
- Gene Name
- magnesium dependent phosphatase 1
- Gene Aliases
- FN6PASE, MDP-1
- Chromosome Location
- Chr. 14: 24213937 - 24216070 on Build GRCh38
- UniGene ID
- Hs.220963
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092821.1 |
- |
4 |
926 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000565988.1 |
- |
5 |
733 |
|
Chr. 14 - 24209646 - 24215974 [-] |
GENCODE |
ENST00000528804.1 |
- |
4 |
926 |
|
Chr. 14 - 24212739 - 24215987 [-] |
View Full Product Details