Gene Transcripts
- Gene Symbol
- ZSCAN30
- Entrez Gene ID
- 100101467
- Gene Name
- zinc finger and SCAN domain containing 30
- Gene Aliases
- ZNF-WYM, ZNF397OS, ZNF917
- Chromosome Location
- Chr. 18: 35251058 - 35290261 on Build GRCh38
- UniGene ID
- Hs.619840
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128697.1 |
- |
2 |
4272 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000586922.2 |
- |
2 |
4276 |
|
Chr. 18 - 35280867 - 35290201 [-] |
View Full Product Details