Gene Transcripts
- Gene Symbol
- DEAR
- Entrez Gene ID
- 102191832
- Gene Name
- dual endothelin-1(VEGFsp)/angiotensin II receptor pseudogene
- Gene Aliases
- DEspR
- Chromosome Location
- Chr. 4: 152337655 - 152338096 on Build GRCh38
- UniGene ID
- Hs.712524
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104273.1 |
|
2 |
262 |
|
GenBank mRNA |
EF212178.1 |
- |
2 |
262 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000605147.1 |
- |
2 |
262 |
|
Chr. 4 - 152337655 - 152338098 [+] |
View Full Product Details