Gene Transcripts
- Gene Symbol
- Drr1
- Entrez Gene ID
- 13499
- Gene Name
- developmentally regulated repeat element-containing transcript 1
- Gene Aliases
- Drr1a, Drr1b, Drr1c
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139652.1 |
- |
2 |
2653 |
|
View Full Product Details