Gene Transcripts
- Gene Symbol
- Fendrr
- Entrez Gene ID
- 68790
- Gene Name
- Foxf1 adjacent non-coding developmental regulatory RNA
- Gene Aliases
- 1110050K14Rik, LL66, linc-Foxf1a, lincFoxf1
- Chromosome Location
- Chr. 8: 121054882 - 121083110 on Build GRCm38
- UniGene ID
- Mm.246916
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004221.1 |
- |
3 |
1066 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB365376.2 |
- |
2 |
669 |
|
Chr. 8 - 121058803 - 121059584 [-] |
View Full Product Details