Gene Transcripts

Gene Symbol
Calcb
Entrez Gene ID
116903
Gene Name
calcitonin-related polypeptide, beta
Gene Aliases
CGRP2, Calc2
Chromosome Location
Chr. 7: 114716556 - 114723368 on Build GRCm38
UniGene ID
Mm.273838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054084.2 NP_473425.2 5 642
GenBank mRNA AK136518.1 - 5 636

View Full Product Details