Gene Transcripts
- Gene Symbol
- PCCA
- Entrez Gene ID
- 5095
- Gene Name
- propionyl-CoA carboxylase alpha subunit
- Gene Aliases
- Chromosome Location
- Chr. 13: 100089015 - 100530437 on Build GRCh38
- UniGene ID
- Hs.733632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000282.3 |
NP_000273.2
|
3, 4 |
337 |
|
NM_001127692.2 |
NP_001121164.1
|
2, 3 |
259 |
|
NM_001178004.1 |
NP_001171475.1
|
3, 4 |
337 |
|
XM_005254059.2 |
XP_005254116.1
|
3, 4 |
332 |
|
XM_011521093.1 |
XP_011519395.1
|
3, 4 |
332 |
|
XM_017020605.1 |
XP_016876094.1
|
3, 4 |
332 |
|
XM_017020606.1 |
XP_016876095.1
|
2, 3 |
183 |
|
XM_017020607.1 |
XP_016876096.1
|
3, 4 |
148 |
|
XM_017020608.1 |
XP_016876097.1
|
3, 4 |
332 |
|
XM_017020609.1 |
XP_016876098.1
|
3, 4 |
245 |
|
XM_017020611.1 |
XP_016876100.1
|
3, 4 |
332 |
|
XM_017020612.1 |
XP_016876101.1
|
3, 4 |
332 |
|
XM_017020613.1 |
XP_016876102.1
|
3, 4 |
332 |
|
XM_017020614.1 |
XP_016876103.1
|
2, 3 |
183 |
|
XM_017020615.1 |
XP_016876104.1
|
3, 4 |
332 |
|
XM_017020616.1 |
XP_016876105.1
|
3, 4 |
332 |
|
GenBank mRNA |
AF385926.1 |
- |
3, 4 |
269 |
|
AK225288.1 |
- |
3, 4 |
237 |
|
AK225299.1 |
- |
3, 4 |
235 |
|
AK296771.1 |
- |
2, 3 |
157 |
|
AK298318.1 |
- |
3, 4 |
259 |
|
AK315503.1 |
- |
3, 4 |
235 |
|
BC000140.2 |
- |
3, 4 |
209 |
3048998 |
DA781564.1 |
- |
2, 3 |
157 |
|
X14608.1 |
- |
3, 4 |
204 |
|
View Full Product Details