Gene Transcripts

Gene Symbol
Vmn1r236
Entrez Gene ID
171235
Gene Name
vomeronasal 1 receptor 236
Gene Aliases
V1rf4
Chromosome Location
Chr. 17: 21286533 - 21287653 on Build GRCm38
UniGene ID
Mm.432089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134201.2 NP_598962.2 1 289
GenBank mRNA BC125459.1 - 1 289
BC125461.1 - 1 289 40129976

View Full Product Details