Gene Transcripts

Gene Symbol
Kcnh6
Entrez Gene ID
192775
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 6
Gene Aliases
m-erg2
Chromosome Location
Chr. 11: 106007994 - 106035595 on Build GRCm38
UniGene ID
Mm.343850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037712.1 NP_001032801.1 11 2354
GenBank mRNA AK038513.1 - 5 853
BC109174.1 - 11 2354 40048550
BC109175.1 - 11 2459

View Full Product Details