Gene Transcripts
- Gene Symbol
- Oog1
- Entrez Gene ID
- 193322
- Gene Name
- oogenesin 1
- Gene Aliases
- Oog, c-1
- Chromosome Location
- Chr. 12: 87552648 - 87608845 on Build GRCm38
- UniGene ID
- Mm.208554
- Species
- Mus musculus
- Gene Symbol
- C87977
- Entrez Gene ID
- 97187
- Gene Name
- expressed sequence C87977
- Gene Aliases
- C85819
- Chromosome Location
- Chr. 4: 144206762 - 144213017 on Build GRCm38
- UniGene ID
- Mm.422698
- Species
- Mus musculus
- Gene Symbol
- Gm10436
- Entrez Gene ID
- 100039315
- Gene Name
- predicted gene 10436
- Gene Aliases
- ENSMUSG00000072907
- Chromosome Location
- Chr. 12: 88175589 - 88210945 on Build GRCm38
- UniGene ID
- Mm.426539
- Species
- Mus musculus
- Gene Symbol
- Gm2042
- Entrez Gene ID
- 100039087
- Gene Name
- predicted gene 2042
- Gene Aliases
- Chromosome Location
- Chr. 12: 87954480 - 87960682 on Build GRCm38
- UniGene ID
- Mm.458830
- Species
- Mus musculus
View Full Product Details