Gene Transcripts

Gene Symbol
Cnksr1
Entrez Gene ID
194231
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
BC037758
Chromosome Location
Chr. 4: 134228041 - 134238416 on Build GRCm38
UniGene ID
Mm.331958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081047.1 NP_001074516.1 15 1421
GenBank mRNA AK145910.1 - 15 1383
BC113805.1 - 15 1386 40062208
BC113956.1 - 15 1386 40062209

View Full Product Details