Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172477.4 | NP_766065.1 | 10 | 2302 | |
GenBank mRNA | AK048041.1 | - | 10 | 2287 | |
AK076686.1 | - | 2 | 445 | ||
AK081176.1 | - | 10 | 2303 | ||
AK135781.1 | - | 8 | 847 | ||
AK160784.1 | - | 10 | 2064 | ||
AK165202.1 | - | 10 | 2157 | ||
AK167298.1 | - | 10 | 2156 | ||
BC056367.1 | - | 10 | 2172 | ||
BC057310.1 | - | 10 | 2172 | ||
BC145599.1 | - | 9 | 1981 | 9054165 | |
BC150799.1 | - | 9 | 1981 | 9087710 |