Gene Transcripts

Gene Symbol
Pgbd5
Entrez Gene ID
209966
Gene Name
piggyBac transposable element derived 5
Gene Aliases
2900019M05Rik, AI854313
Chromosome Location
Chr. 8: 124369049 - 124434462 on Build GRCm38
UniGene ID
Mm.234233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_171824.2 NP_741958.1 6 1170
GenBank mRNA BC094384.1 - 1168

View Full Product Details