Gene Transcripts

Gene Symbol
Arfgef1
Entrez Gene ID
211673
Gene Name
ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited)
Gene Aliases
ARFGEP1, BIG1, D130059B05Rik, D730028O18Rik, P200
Chromosome Location
Chr. 1: 10137507 - 10232670 on Build GRCm38
UniGene ID
Mm.229141
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102430.1 NP_001095900.1 26 3940
GenBank mRNA AK134369.1 - 2 214

View Full Product Details