Gene Transcripts

Gene Symbol
Sprn
Entrez Gene ID
212518
Gene Name
shadow of prion protein
Gene Aliases
C630041J07
Chromosome Location
Chr. 7: 140150628 - 140154659 on Build GRCm38
UniGene ID
Mm.246858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183147.2 NP_898970.1 2 1107
GenBank mRNA AK049995.1 - 2 1144
BC056484.1 - 2 1107 5696462

View Full Product Details