Gene Transcripts

Gene Symbol
Aldh4a1
Entrez Gene ID
212647
Gene Name
aldehyde dehydrogenase 4 family, member A1
Gene Aliases
A930035F14Rik, Ahd-1, Ahd1, Aldh4, Aldh5a1, E330022C09, P5cd, P5cdh, P5cdhl, P5cdhs, Ssdh1
Chromosome Location
Chr. 4: 139622871 - 139649691 on Build GRCm38
UniGene ID
Mm.273571
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175438.4 NP_780647.3 12 1534
GenBank mRNA AK044712.1 - 12 1330
BC024133.1 - 11 1252
BC026589.1 - 7 746
BC027140.1 - 8 946
BC039280.1 - 1 98
BC039281.1 - 12 1318 4975491
BC056226.1 - 12 1418 5064715
BC060650.1 - 11 2924 30354228
CD354968.1 - 2 166

View Full Product Details