Gene Transcripts

Gene Symbol
Gm44
Entrez Gene ID
212952
Gene Name
predicted gene 44
Gene Aliases
Chromosome Location
Chr. X: 90878355 - 90893134 on Build GRCm38
UniGene ID
Mm.477436
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101450.1 NP_001094920.1 1 471

View Full Product Details