Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178674.4 | NP_848789.2 | 4 | 514 | |
GenBank mRNA | AK035290.1 | - | 3 | 402 | |
AK046196.1 | - | 4 | 510 | ||
AK079800.1 | - | 3 | 420 | ||
AK085598.1 | - | 4 | 515 | ||
BC138940.1 | - | 4 | 492 | 8861962 | |
BC138941.1 | - | 4 | 492 | 8861963 |