Gene Transcripts

Gene Symbol
Lemd1
Entrez Gene ID
213409
Gene Name
LEM domain containing 1
Gene Aliases
4930540I23Rik, 6430514M23Rik, AI428128, LEMP-1
Chromosome Location
Chr. 1: 132190687 - 132257382 on Build GRCm38
UniGene ID
Mm.426321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033250.4 NP_001028422.2 4 352
GenBank mRNA AK016010.1 - 5 497
AK144269.1 - 430
BC115462.1 - 4 333 40100287
BC115463.1 - 4 333 40100292

View Full Product Details