Gene Transcripts

Gene Symbol
Slc16a11
Entrez Gene ID
216867
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 11
Gene Aliases
AB041591, MNCb-2717
Chromosome Location
Chr. 11: 70213910 - 70216414 on Build GRCm38
UniGene ID
Mm.289238
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153081.3 NP_694721.2 4 1086
GenBank mRNA AB041591.1 - 4 879
BC130255.1 - 3 589

View Full Product Details