Gene Transcripts
- Gene Symbol
- PGR
- Entrez Gene ID
- 5241
- Gene Name
- progesterone receptor
- Gene Aliases
- NR3C3, PR
- Chromosome Location
- Chr. 11: 101029624 - 101130681 on Build GRCh38
- UniGene ID
- Hs.32405
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000926.4 |
NP_000917.3
|
4, 5 |
2963 |
|
NM_001202474.3 |
NP_001189403.1
|
4, 5 |
2213 |
|
NM_001271162.1 |
NP_001258091.1
|
4, 5 |
883 |
|
NR_073141.2 |
|
4, 5 |
2213 |
|
NR_073142.2 |
|
3, 4 |
2096 |
|
XM_006718858.3 |
XP_006718921.1
|
4, 5 |
3813 |
|
GenBank mRNA |
AB085843.1 |
- |
3, 4 |
2103 |
|
AB085845.1 |
- |
4, 5 |
2220 |
|
AF016381.1 |
- |
4, 5 |
2395 |
|
AK304853.1 |
- |
4, 5 |
883 |
|
AY212933.1 |
- |
1, 2 |
1593 |
|
AY382151.1 |
- |
3, 4 |
579 |
|
M15716.1 |
- |
4, 5 |
2395 |
|
X51730.1 |
- |
4, 5 |
3674 |
|
View Full Product Details